siRNA Details
| Gene Symbol: Klhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290477.1 | NP_001277406.1 | 3, 4 | 807 | ||
| NM_001290478.1 | NP_001277407.1 | 3, 4 | 807 | |||
| NM_172781.2 | NP_766369.2 | 3, 4 | 807 | |||
| GenBank mRNA | AK044930.1 | - | 3, 4 | 538 | ||
| AK081448.1 | - | 3, 4 | 808 | |||
| AK081937.1 | - | 3, 4 | 620 | |||
| AK085099.1 | - | 3, 4 | 808 | |||
| AK161786.1 | - | 3, 4 | 701 | |||
| BC057137.1 | - | 3, 4 | 538 | |||
| BC099954.1 | - | 3, 4 | 634 | 30081872 | ||
| BC141251.1 | - | 3, 4 | 550 | 9055863 | ||

Klhl4
Genomic Map