siRNA Details
| Gene Symbol: Klhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290477.1 | NP_001277406.1 | 8 | 1672 | ||
| NM_001290478.1 | NP_001277407.1 | 8 | 1591 | |||
| NM_172781.2 | NP_766369.2 | 8 | 1690 | |||
| GenBank mRNA | AK044930.1 | - | 8 | 1322 | ||
| AK081448.1 | - | 8 | 1674 | |||
| AK081937.1 | - | 8 | 1503 | |||
| AK085099.1 | - | 8 | 1673 | |||
| AK161786.1 | - | 8 | 1566 | |||
| BC046395.1 | - | 2 | 250 | |||
| BC057137.1 | - | 8 | 1421 | |||
| BC141251.1 | - | 8 | 1415 | 9055863 | ||

Klhl4
Genomic Map