siRNA Details
| Gene Symbol: 4932411N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177705.3 | NP_808373.1 | 3 | 1720 | ||
| GenBank mRNA | AK029976.1 | - | 3 | 1720 | ||
| BC117864.1 | - | 1 | 1326 | 40095418 | ||
| BC117865.1 | - | 1 | 1326 | 40095419 | ||

4932411N23Rik
Genomic Map