siRNA Details
| Gene Symbol: Rbm41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172147.1 | NP_001165618.1 | 7 | 1109 | ||
| NM_001172148.1 | NP_001165619.1 | 6 | 1027 | |||
| NM_001290630.1 | NP_001277559.1 | 7 | 1099 | |||
| NM_153586.2 | NP_705814.1 | 6 | 1027 | |||
| GenBank mRNA | AK084514.1 | - | 6 | 962 | ||
| AK084629.1 | - | 6 | 962 | |||
| AK141075.1 | - | 7 | 1034 | |||
| AK163147.1 | - | 5 | 947 | |||
| AK164496.1 | - | 7 | 1057 | |||
| AK166005.1 | - | 6 | 962 | |||
| BC037024.1 | - | 6 | 994 | 5368185 | ||

Rbm41
Genomic Map