siRNA Details
Gene Symbol: Adgrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079847.2 | NP_001073316.1 | 17 | 1513 | ||
NM_001079848.2 | NP_001073317.1 | 16 | 1483 | |||
NM_001079857.2 | NP_001073326.1 | 17 | 1522 | |||
NM_001290445.1 | NP_001277374.1 | 17 | 1516 | |||
NM_001290446.1 | NP_001277375.1 | 18 | 1555 | |||
NM_178712.4 | NP_848827.1 | 18 | 1564 | |||
GenBank mRNA | AF538952.1 | - | 18 | 1529 | ||
AF538955.1 | - | 17 | 1481 | |||
AF538956.1 | - | 18 | 1520 | |||
AF538957.1 | - | 16 | 1448 | |||
AK041291.1 | - | 19 | 1752 | |||
AK046871.1 | - | 17 | 1489 | |||
BC115874.1 | - | 16 | 1458 | 40086356 | ||
BC116644.1 | - | 17 | 1509 | 40086357 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
