siRNA Details
| Gene Symbol: Fancb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146081.1 | NP_001139553.1 | 5 | 1523 | ||
| NM_175027.4 | NP_778192.3 | 5 | 1523 | |||
| GenBank mRNA | AK040957.1 | - | 1 | 626 | ||
| AK132251.1 | - | 5 | 1523 | |||
| AK169374.1 | - | 5 | 1411 | |||
| BC083151.1 | - | 5 | 1416 | 30460698 | ||

Fancb
Genomic Map