siRNA Details
| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172785.3 | NP_766373.2 | 5 | 901 | ||
| GenBank mRNA | AK052806.1 | - | 5 | 903 | ||
| BC112398.1 | - | 4 | 683 | 40062049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h12d| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172785.3 | NP_766373.2 | 5 | 901 | ||
| GenBank mRNA | AK052806.1 | - | 5 | 903 | ||
| BC112398.1 | - | 4 | 683 | 40062049 | ||