siRNA Details
| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178713.4 | NP_848828.1 | 7 | 1176 | ||
| GenBank mRNA | AF510322.1 | - | 7 | 1185 | ||
| AK050298.1 | - | 7 | 1177 | |||
| AK143752.1 | - | 7 | 1177 | |||
| BC013511.1 | - | 5 | 718 | |||
| BC038493.1 | - | 7 | 1188 | 4223059 | ||

Aldh8a1
Genomic Map