siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172787.2 | NP_766375.1 | 12, 13 | 1336 | ||
| GenBank mRNA | AK045667.1 | - | 12, 13 | 1337 | ||
| AK049597.1 | - | 12, 13 | 1371 | |||
| AK129443.1 | - | 8, 9 | 881 | |||
| BC082309.1 | - | 11, 12 | 1206 | 30356672 | ||
| BC085192.1 | - | 11, 12 | 1441 | 30694487 | ||

L3mbtl3
Genomic Map