siRNA Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081127.1 | NP_001074596.1 | 12 | 2028 | ||
| GenBank mRNA | AK145103.1 | - | 12 | 2030 | ||
| BC157953.1 | - | 12 | 1863 | 9007283 | ||
| BC158014.1 | - | 12 | 1861 | 9088151 | ||
| BC172106.1 | - | 12 | 1858 | |||

Adamts14
Genomic Map