siRNA Details
| Gene Symbol: Mex3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198615.2 | NP_941017.2 | 1 | 894 | ||
| GenBank mRNA | AK149801.1 | - | 1 | 422 | ||
| BC059858.1 | - | 1 | 70 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mex3d| Gene Symbol: Mex3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198615.2 | NP_941017.2 | 1 | 894 | ||
| GenBank mRNA | AK149801.1 | - | 1 | 422 | ||
| BC059858.1 | - | 1 | 70 | |||