siRNA Details
Gene Symbol: Lingo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013758.2 | NP_001013780.2 | 2 | 1334 | ||
GenBank mRNA | BC072620.1 | - | 2 | 1325 | 6815693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lingo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013758.2 | NP_001013780.2 | 2 | 1334 | ||
GenBank mRNA | BC072620.1 | - | 2 | 1325 | 6815693 |