siRNA Details
| Gene Symbol: Zfp938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105557.2 | NP_001099027.2 | 4 | 1158 | ||
| GenBank mRNA | BC040349.1 | - | 4 | 1164 | 4024367 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp938| Gene Symbol: Zfp938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105557.2 | NP_001099027.2 | 4 | 1158 | ||
| GenBank mRNA | BC040349.1 | - | 4 | 1164 | 4024367 | |