siRNA Details
| Gene Symbol: Gas2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033331.2 | NP_001028503.2 | 11 | 1393 | ||
| NM_001079876.1 | NP_001073345.1 | 11 | 1396 | |||
| GenBank mRNA | AK136054.1 | - | 11 | 1406 | ||
| AK170634.1 | - | 11 | 1328 | |||
| AK170750.1 | - | 11 | 1318 | |||

Gas2l3
Genomic Map