siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 6, 7 | 953 | ||
| GenBank mRNA | AK078325.1 | - | 6, 7 | 954 | ||
| BC031778.1 | - | 500 | 4501115 | |||
| BC049904.1 | - | 500 | 4501115 | |||
| BC064815.1 | - | 6, 7 | 938 | 6516981 | ||
| BY130550.1 | - | 2, 3 | 177 | |||

Cdk17
Genomic Map