siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 9 | 1207 | ||
| GenBank mRNA | BC031778.1 | - | 754 | 4501115 | ||
| BC049904.1 | - | 754 | 4501115 | |||
| BC064815.1 | - | 9 | 1192 | 6516981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk17| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 9 | 1207 | ||
| GenBank mRNA | BC031778.1 | - | 754 | 4501115 | ||
| BC049904.1 | - | 754 | 4501115 | |||
| BC064815.1 | - | 9 | 1192 | 6516981 | ||