siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 12 | 1556 | ||
| GenBank mRNA | BC031778.1 | - | 1103 | 4501115 | ||
| BC049904.1 | - | 1103 | 4501115 | |||
| BC064815.1 | - | 12 | 1541 | 6516981 | ||
| BQ442862.1 | - | 2 | 140 | |||

Cdk17
Genomic Map