siRNA Details
| Gene Symbol: Tmtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033332.2 | NP_001028504.1 | 2 | 324 | ||
| NM_001110013.1 | NP_001103483.1 | 2 | 324 | |||
| GenBank mRNA | AK044857.1 | - | 2 | 280 | ||
| AK146769.1 | - | 2 | 317 | |||
| AK149759.1 | - | 2 | 283 | |||
| AK165838.1 | - | 2 | 325 | |||

Tmtc3
Genomic Map