siRNA Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146241.2 | NP_666353.1 | 13 | 2354 | ||
| GenBank mRNA | AK029833.1 | - | 1684 | |||
| AK034093.1 | - | 13 | 1733 | |||
| BC032288.1 | - | 13 | 2359 | 5368580 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108991, XM_008765360, X80535.

Trhde
Genomic Map