siRNA Details
| Gene Symbol: Myrfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033333.3 | NP_001028505.1 | 6 | 889 | ||
| GenBank mRNA | AK144383.1 | - | 6 | 890 | ||
| AK144411.1 | - | 6 | 890 | |||
| BC150915.1 | - | 6 | 693 | 9087826 | ||
| BC150923.1 | - | 6 | 693 | 9087834 | ||

Myrfl
Genomic Map