siRNA Details
| Gene Symbol: Ankrd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172790.2 | NP_766378.1 | 17 | 1841 | ||
| GenBank mRNA | AK090124.1 | - | 17 | 1841 | ||
| BC117907.1 | - | 17 | 1792 | 40097189 | ||
| BC117908.1 | - | 17 | 1792 | 40097194 | ||

Ankrd52
Genomic Map