siRNA Details
| Gene Symbol: Pla2g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172791.2 | NP_766379.2 | 7 | 1560 | ||
| GenBank mRNA | AK033572.1 | - | 7 | 1504 | ||
| AK046215.1 | - | 4 | 1674 | |||
| BC079556.1 | - | 7 | 1560 | 30360278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g3| Gene Symbol: Pla2g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172791.2 | NP_766379.2 | 7 | 1560 | ||
| GenBank mRNA | AK033572.1 | - | 7 | 1504 | ||
| AK046215.1 | - | 4 | 1674 | |||
| BC079556.1 | - | 7 | 1560 | 30360278 | ||