siRNA Details
| Gene Symbol: Zfp454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172794.2 | NP_766382.2 | 6 | 1540 | ||
| GenBank mRNA | AK046117.1 | - | 3 | 1289 | ||
| BC049793.1 | - | 6 | 1540 | 6510760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp454| Gene Symbol: Zfp454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172794.2 | NP_766382.2 | 6 | 1540 | ||
| GenBank mRNA | AK046117.1 | - | 3 | 1289 | ||
| BC049793.1 | - | 6 | 1540 | 6510760 | ||