siRNA Details
| Gene Symbol: Sowaha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183173.2 | NP_898996.2 | 1 | 708 | ||
| GenBank mRNA | AK043538.1 | - | 1 | 595 | ||
| BC117862.1 | - | 1 | 46 | |||
| BC117863.1 | - | 1 | 46 | 40095300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sowaha| Gene Symbol: Sowaha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183173.2 | NP_898996.2 | 1 | 708 | ||
| GenBank mRNA | AK043538.1 | - | 1 | 595 | ||
| BC117862.1 | - | 1 | 46 | |||
| BC117863.1 | - | 1 | 46 | 40095300 | ||