siRNA Details
| Gene Symbol: Gm12258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CN460644.1 | - | 5 | 423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12258| Gene Symbol: Gm12258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CN460644.1 | - | 5 | 423 | ||