siRNA Details
| Gene Symbol: Mief2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009927.2 | NP_001009927.1 | 4 | 524 | ||
| GenBank mRNA | AK156917.1 | - | 4 | 507 | ||
| AK171444.1 | - | 3 | 649 | |||
| BC088984.1 | - | 4 | 413 | 5708020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mief2| Gene Symbol: Mief2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009927.2 | NP_001009927.1 | 4 | 524 | ||
| GenBank mRNA | AK156917.1 | - | 4 | 507 | ||
| AK171444.1 | - | 3 | 649 | |||
| BC088984.1 | - | 4 | 413 | 5708020 | ||