siRNA Details
| Gene Symbol: Krt39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213730.2 | NP_998895.2 | 4, 5 | 866 | ||
| GenBank mRNA | BC119541.1 | - | 4, 5 | 866 | ||
| BC128563.1 | - | 4, 5 | 861 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt39| Gene Symbol: Krt39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213730.2 | NP_998895.2 | 4, 5 | 866 | ||
| GenBank mRNA | BC119541.1 | - | 4, 5 | 866 | ||
| BC128563.1 | - | 4, 5 | 861 | |||