siRNA Details
| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 3 | 365 | ||
| GenBank mRNA | AK030591.1 | - | 1463 | |||
| AK052040.1 | - | 3 | 390 | |||
| AY351699.1 | - | 3 | 365 | |||
| BC038036.1 | - | 3 | 643 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdk2| Gene Symbol: Sdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172800.2 | NP_766388.2 | 3 | 365 | ||
| GenBank mRNA | AK030591.1 | - | 1463 | |||
| AK052040.1 | - | 3 | 390 | |||
| AY351699.1 | - | 3 | 365 | |||
| BC038036.1 | - | 3 | 643 | |||