siRNA Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001333.2 | NP_001001333.2 | 5 | 852 | ||
| NM_001146073.1 | NP_001139545.1 | 4 | 438 | |||
| GenBank mRNA | AK154237.1 | - | 5 | 826 | ||
| BC056487.1 | - | 5 | 2433 | |||
| BC069960.1 | - | 4 | 390 | 5007884 | ||
| EF437388.1 | - | 11 | ||||
| FM204885.1 | - | 4 | 389 | |||
| FM204886.1 | - | 4 | 389 | |||

Hexdc
Genomic Map