siRNA Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146073.1 | NP_001139545.1 | 9 | 1200 | ||
| GenBank mRNA | BC056487.1 | - | 8 | 2928 | ||
| BC069960.1 | - | 9 | 1152 | 5007884 | ||
| FM204886.1 | - | 9 | 1151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hexdc| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146073.1 | NP_001139545.1 | 9 | 1200 | ||
| GenBank mRNA | BC056487.1 | - | 8 | 2928 | ||
| BC069960.1 | - | 9 | 1152 | 5007884 | ||
| FM204886.1 | - | 9 | 1151 | |||