siRNA Details
Gene Symbol: Gdf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013527.1 | NP_038555.1 | 2 | 1306 | ||
GenBank mRNA | AF525752.1 | - | 2 | 1306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gdf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013527.1 | NP_038555.1 | 2 | 1306 | ||
GenBank mRNA | AF525752.1 | - | 2 | 1306 |