siRNA Details
| Gene Symbol: Dock4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172803.2 | NP_766391.2 | 34 | 3818 | ||
| GenBank mRNA | AK047749.1 | - | 3 | 219 | ||
| AK049953.1 | - | 4 | 984 | |||
| AK122353.1 | - | 34 | 3800 | |||
| BC051125.1 | - | 2 | 116 | |||
| BC141138.1 | - | 34 | 3535 | 9055750 | ||

Dock4
Genomic Map