siRNA Details
| Gene Symbol: Vash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177354.4 | NP_796328.2 | 2 | 1643 | ||
| GenBank mRNA | AB284948.1 | - | 2 | 1648 | ||
| AK090166.1 | - | 2 | 1649 | |||
| BC086688.1 | - | 2 | 143 | 30686635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vash1| Gene Symbol: Vash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177354.4 | NP_796328.2 | 2 | 1643 | ||
| GenBank mRNA | AB284948.1 | - | 2 | 1648 | ||
| AK090166.1 | - | 2 | 1649 | |||
| BC086688.1 | - | 2 | 143 | 30686635 | ||