siRNA Details
| Gene Symbol: Cntnap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081129.1 | NP_001074598.1 | 17 | 2664 | ||
| GenBank mRNA | AK141609.1 | - | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntnap3| Gene Symbol: Cntnap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081129.1 | NP_001074598.1 | 17 | 2664 | ||
| GenBank mRNA | AK141609.1 | - | 287 | |||