siRNA Details
| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 510 | ||
| GenBank mRNA | AK133308.1 | - | 511 | |||
| AK161458.1 | - | 503 | ||||
| BC062958.1 | - | 4 | 510 | 30548236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp458| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 510 | ||
| GenBank mRNA | AK133308.1 | - | 511 | |||
| AK161458.1 | - | 503 | ||||
| BC062958.1 | - | 4 | 510 | 30548236 | ||