siRNA Details
| Gene Symbol: Zfp366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004149.1 | NP_001004149.1 | 5 | 2059 | ||
| GenBank mRNA | BC070399.1 | - | 5 | 2059 | 5694313 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp366| Gene Symbol: Zfp366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004149.1 | NP_001004149.1 | 5 | 2059 | ||
| GenBank mRNA | BC070399.1 | - | 5 | 2059 | 5694313 | |