siRNA Details
| Gene Symbol: Fam160b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194345.1 | NP_919326.1 | 4 | 370 | ||
| GenBank mRNA | BC050861.1 | - | 4 | 371 | ||
| BU152183.1 | - | 4 | 370 | |||
| CF170130.1 | - | 4 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam160b2| Gene Symbol: Fam160b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194345.1 | NP_919326.1 | 4 | 370 | ||
| GenBank mRNA | BC050861.1 | - | 4 | 371 | ||
| BU152183.1 | - | 4 | 370 | |||
| CF170130.1 | - | 4 | 326 | |||