siRNA Details
| Gene Symbol: Kctd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177715.4 | NP_808383.3 | 1 | 1673 | ||
| GenBank mRNA | AK035535.1 | - | 1 | 1675 | ||
| AK087895.1 | - | 1 | 1681 | |||
| AK172581.1 | - | 1 | 1674 | |||
| AK173260.1 | - | 1 | 1178 | |||
| BC116647.1 | - | 2 | 1060 | |||
| BC116648.1 | - | 2 | 1060 | |||

Kctd12
Genomic Map