siRNA Details
Gene Symbol: Rxfp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178717.3 | NP_848832.1 | 1 | 1585 | ||
GenBank mRNA | AK043414.1 | - | 1 | 1610 | ||
AK046367.1 | - | 1 | 1336 | |||
AK084001.1 | - | 1 | 1587 | |||
AY170825.1 | - | 1 | 901 | |||
AY633762.1 | - | 1 | 901 | |||
BC053073.1 | - | 1 | 1007 | 5686363 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
