siRNA Details
| Gene Symbol: Adamts12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175501.2 | NP_780710.2 | 19 | 3566 | ||
| GenBank mRNA | AJ537452.1 | - | 19 | 3325 | ||
| AK028459.1 | - | 10 | 1821 | |||
| BC151248.1 | - | 10 | 1690 | |||
| BC152338.1 | - | 10 | 1690 | |||

Adamts12
Genomic Map