siRNA Details
| Gene Symbol: Tspyl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085421.1 | NP_001078890.1 | 1 | 1162 | ||
| GenBank mRNA | AK031988.1 | - | 1 | 873 | ||
| AK087510.1 | - | 1 | 1098 | |||
| AK173253.1 | - | 1 | 1105 | |||
| BC079547.1 | - | 1 | 1113 | |||

Tspyl5
Genomic Map