siRNA Details
| Gene Symbol: Erich5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173421.2 | NP_775597.1 | 2 | 429 | ||
| GenBank mRNA | AK135792.1 | - | 2 | 160 | ||
| AK162262.1 | - | 2 | 187 | |||
| BC030476.1 | - | 2 | 437 | 5368170 | ||
| BC099414.1 | - | 2 | 180 | 30931867 | ||

Erich5
Genomic Map