siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172815.3 | NP_766403.1 | 3, 4 | 1164 | ||
| GenBank mRNA | AK049891.1 | - | 3, 4 | 915 | ||
| AK052457.1 | - | 2, 3 | 1288 | |||
| AK087485.1 | - | 3, 4 | 1165 | |||
| BC052844.1 | - | 3, 4 | 515 | |||

Rspo2
Genomic Map