siRNA Details
| Gene Symbol: Tmem74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175502.3 | NP_780711.1 | 2 | 604 | ||
| GenBank mRNA | AK045145.1 | - | 612 | |||
| AK046420.1 | - | 2 | 605 | |||
| BC076589.1 | - | 599 | ||||
| BC117021.1 | - | 1 | 196 | 40126340 | ||
| BC117025.1 | - | 1 | 196 | 40126344 | ||

Tmem74
Genomic Map