siRNA Details
| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173862.2 | NP_776287.2 | 1 | 574 | ||
| GenBank mRNA | BC030396.1 | - | 1 | 577 | 4487327 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83a| Gene Symbol: Fam83a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173862.2 | NP_776287.2 | 1 | 574 | ||
| GenBank mRNA | BC030396.1 | - | 1 | 577 | 4487327 | |