siRNA Details
Gene Symbol: Foxred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017983.2 | NP_001017983.2 | 2 | 300 | ||
NM_001168260.1 | NP_001161732.1 | 2 | 398 | |||
GenBank mRNA | AK140027.1 | - | 2 | 260 | ||
AK170795.1 | - | 2 | 353 | |||
BC086662.1 | - | 2 | 230 | 30549783 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
