siRNA Details
| Gene Symbol: Ttc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033337.4 | NP_001028509.2 | 7, 8 | 823 | ||
| GenBank mRNA | AK078848.1 | - | 7, 8 | 784 | ||
| AK133636.1 | - | 6, 7 | 531 | |||
| AK200042.1 | - | 1, 2 | 75 | |||
| AK204138.1 | - | 1, 2 | 75 | |||
| BC024550.1 | - | 7, 8 | 773 | |||
| BC030849.1 | - | 7, 8 | 786 | |||
| BC132621.1 | - | 7, 8 | 808 | 40130898 | ||
| BC132623.1 | - | 7, 8 | 808 | 40130900 | ||

Ttc38
Genomic Map