siRNA Details
| Gene Symbol: Ttll8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172818.3 | NP_766406.2 | 12 | 2550 | ||
| GenBank mRNA | AK030151.1 | - | 12 | 2543 | ||
| AM690752.1 | - | 12 | 2207 | |||
| BC116293.1 | - | 12 | 2191 | 40094952 | ||
| BC116294.1 | - | 12 | 2186 | 40094955 | ||

Ttll8
Genomic Map