siRNA Details
| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033633.3 | NP_001028805.2 | 9 | 1728 | ||
| GenBank mRNA | AK147341.1 | - | 9 | 1729 | ||
| BC119586.1 | - | 10 | 1295 | 40058865 | ||
| BC119587.1 | - | 9 | 1237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a13| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033633.3 | NP_001028805.2 | 9 | 1728 | ||
| GenBank mRNA | AK147341.1 | - | 9 | 1729 | ||
| BC119586.1 | - | 10 | 1295 | 40058865 | ||
| BC119587.1 | - | 9 | 1237 | |||