siRNA Details
| Gene Symbol: Dip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159361.1 | NP_001152833.1 | 19 | 2383 | ||
| NM_172819.3 | NP_766407.2 | 14 | 1595 | |||
| GenBank mRNA | AB093297.1 | - | 4 | 399 | ||
| AK030064.1 | - | 14 | 1596 | |||
| AK078824.1 | - | 4 | 419 | |||
| AK147562.1 | - | 19 | 2319 | |||
| BC023757.1 | - | 5 | 482 | |||
| BC137857.1 | - | 14 | 1582 | 8860876 | ||
| BC137858.1 | - | 14 | 1582 | 8860877 | ||

Dip2b
Genomic Map